Learn More
Abcam LY2874455, FGF/FGFR inhibitor, 5MG
Catalog No. 502676654
Encompass_Preferred
MW 444.3 Da, Purity >98%. Potent FGF/FGFR inhibitor. Inhibits in vitro autophosphorylation of FGFR-1, FGFR-2, FGFR-3, and FGFR-4 with similar inhibition potency for all four FGFRs (IC₅₀ < 6.4 nM). Exhibits a 6- to 9-fold in vitro and in vivo selectivity on inhibition of FGF- over VEGF-mediated target signaling in mice.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6654 Supplier Abcam Supplier No. AB2163135MG
MW 444.3 Da, Purity >98%. Potent FGF/FGFR inhibitor. Inhibits in vitro autophosphorylation of FGFR-1, FGFR-2, FGFR-3, and FGFR-4 with similar inhibition potency for all four FGFRs (IC₅₀ < 6.4 nM). Exhibits a 6- to 9-fold in vitro and in vivo selectivity on inhibition of FGF- over VEGF-mediated target signaling in mice.
The product is subject to the following: Abcam Restricted Use Statement