Learn More
Apexbio Technology LLC LY2886721 1262036-50-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A84655.1
Encompass_Preferred
LY2886721 (CAS 1262036-50-9) is an orally bioavailable inhibitor of -site amyloid precursor protein cleaving enzyme 1 (BACE1) an aspartyl protease integral to amyloid precursor protein (APP) cleavage and amyloid- (A ) generation LY2886721 inhibits BACE1 activity with an IC50 of approximately 20 3 nM effectively reducing soluble APP and elevating soluble APP in human cell models (HEK293Swe) and neuronal cultures (PDAPP) at IC50 values of 18 7 nM and 10 7 nM respectively In animal models such as PDAPP mice oral LY2886721 administration dose-dependently reduces cerebral A C99 and sAPP levels Due to its BACE1 inhibitory action LY2886721 serves as a valuable tool in Alzheimer s disease research and therapeutic development
Catalog No. 50-000-61682