Learn More
Abcam LY2886721, BACE-1 inhibitor, 5MG
Catalog No. 502702879
Encompass_Preferred
MW 390.4 Da, Purity >98%. Potent and selective BACE-1 inhibitor (IC₅₀ = 20.3 nM for recombinant hBACE-1). Human BACE2 is inhibited with an IC₅₀ of 10.2 nM. Does not inhibit other aspartyl proteases such as cathepsin D, pepsin, and renin. A potential agent to treat Alzheimer's Disease. Inhibits production of Aβ in HEK-293 Swe cells (IC₅₀ values are 18.5 nM and 19.7 nM for Aβ1-40 and Aβ1-42 respectively) and in PDAPP neuronal cells (IC₅₀ ~10 nM). Reduces dose-dependently brain levels of Aβ, C99 and sAPPβ in orally administered mouse model and plasma Aβ in dog model. Ppenetrates the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-2879 Supplier Abcam Supplier No. AB2238865MG
MW 390.4 Da, Purity >98%. Potent and selective BACE-1 inhibitor (IC₅₀ = 20.3 nM for recombinant hBACE-1). Human BACE2 is inhibited with an IC₅₀ of 10.2 nM. Does not inhibit other aspartyl proteases such as cathepsin D, pepsin, and renin. A potential agent to treat Alzheimer's Disease. Inhibits production of Aβ in HEK-293 Swe cells (IC₅₀ values are 18.5 nM and 19.7 nM for Aβ1-40 and Aβ1-42 respectively) and in PDAPP neuronal cells (IC₅₀ ~10 nM). Reduces dose-dependently brain levels of Aβ, C99 and sAPPβ in orally administered mouse model and plasma Aβ in dog model. Ppenetrates the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement