Learn More
Selleck Chemical LLC LY2886721 50mg 1262036-50-9
SDP
Supplier:  Selleck Chemical LLC S215650MG
Encompass_Preferred
LY2886721 is a BACE inhibitor used for the treatment of Alzheimer's Disease. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-773-9