Learn More
Medchemexpress LLC HY-19835 5mg Medchemexpress, LY2922470 CAS:1423018-12-5 Purity:>98%
SDP
Catalog No. 501970038
Encompass_Preferred
Medchemexpress, HY-19835 5mg LY2922470 CAS:1423018-12-5 LY2922470 is a potent, selective and orally available agonist of the G protein-coupled receptor 40 (GPR40), with EC50s of 7 nM, 1 nM and 3 nM for human GPR40, mouse GPR40 and rat GPR40, respectively. LY2922470 reduces glucose levels along with significant increases in insulin and GLP-1, is potential for the treatment of type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0038 Supplier Medchemexpress LLC Supplier No. HY198355MG
Medchemexpress, HY-19835 5mg LY2922470 CAS:1423018-12-5 LY2922470 is a potent, selective and orally available agonist of the G protein-coupled receptor 40 (GPR40), with EC50s of 7 nM, 1 nM and 3 nM for human GPR40, mouse GPR40 and rat GPR40, respectively. LY2922470 reduces glucose levels along with significant increases in insulin and GLP-1, is potential for the treatment of type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.