Learn More
Medchemexpress LLC HY-10108 500mg , LY294002 NSC 697286;SF 1101 CAS:154447-36-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10108500MG
Encompass_Preferred
Medchemexpress, HY-10108 500mg LY294002 NSC 697286;SF 1101 CAS:154447-36-6 Formula:C19H17NO3 IC50: 0.5/0.57/0.97 μM (PI3Kα/δ/β) IC50: 98 nM (CK2) Purity:98% LY294002 is a broad-spectrum inhibitor of PI3K , with IC 50 of 0.5/0.57/0.97 μM for PI3Kα / PI3Kδ / PI3Kβ , respectively, also potently inhibits CK2 with IC 50 of 98 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2334