Learn More
Medchemexpress LLC HY-10108 50mg Medchemexpress, LY294002 CAS:154447-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010850MG
Encompass_Preferred
Medchemexpress, HY-10108 50mg LY294002 CAS:154447-36-6 LY294002 is a broad-spectrum inhibitor of PI3K with IC50s of 0.5, 0.57, and 0.97 μM for PI3Kα, PI3Kδ and PI3Kβ, respectively. LY294002 also inhibits CK2 with an IC50 of 98 nM[2]. LY294002 is a competitive DNA-PK inhibitorr that binds reversibly to the kinase domain of DNA-PK with an IC50 of 1.4 μM. LY294002 is an autophagy and apoptosis activator[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8645