Learn More
Selleck Chemical LLC LY294002 200mg 154447-36-6 SF 1101, NSC 697286
SDP
Catalog No. 508537
Encompass_Preferred
LY294002 (SF 1101, NSC 697286) is the first synthetic molecule known to inhibit PI3K?/?/? with IC50 of 0.5 ?M/0.57 ?M/0.97 ?M, respectively; more stable in solution than Wortmannin, and also blocks autophagosome formation. It not only binds to class I PI3Ks and other PI3K-related kinases, but also to novel targets seemingly unrelated to the PI3K family. LY294002 also inhibits CK2 with IC50 of 98 nM. LY294002 is a non-specific DNA-PKcs inhibitor and activates autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-853-7 Supplier Selleck Chemical LLC Supplier No. S1105200MG
LY294002 (SF 1101, NSC 697286) is the first synthetic molecule known to inhibit PI3K?/?/? with IC50 of 0.5 ?M/0.57 ?M/0.97 ?M, respectively; more stable in solution than Wortmannin, and also blocks autophagosome formation. It not only binds to class I PI3Ks and other PI3K-related kinases, but also to novel targets seemingly unrelated to the PI3K family. LY294002 also inhibits CK2 with IC50 of 98 nM. LY294002 is a non-specific DNA-PKcs inhibitor and activates autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.