Learn More
Echelon Biosciences Research Labs LY294002 (HCI SALT) 25 mg
SDP
Catalog No. 50720010
Encompass
LY294002 is a selective, cell permeable, potent and specific inhibitor of phosphatidylinositol 3-kinase (IC50 = 1.4 M; purified PI 3-kinase). It does not significantly inhibit PKC, PKA, MAP kinase, S6 kinase, EGFrK, c-src kinase, PI 4-kinase, DAG kinase or rabbit kidney ATPase at concentrations of 50 M. Induces apoptosis. In intact neutrophils stimulated with fMet-Leu-Phe, LY294002 completely abolished PI 3-kinase activity at 50 M without cell toxicity.nn
nn
Specifications
nProvided as HCl saltnNot Sterilen
Featured in Publications
This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-010 Supplier Echelon Biosciences Research Labs Supplier No. B029425MG
LY294002 is a selective, cell permeable, potent and specific inhibitor of phosphatidylinositol 3-kinase (IC50 = 1.4 M; purified PI 3-kinase). It does not significantly inhibit PKC, PKA, MAP kinase, S6 kinase, EGFrK, c-src kinase, PI 4-kinase, DAG kinase or rabbit kidney ATPase at concentrations of 50 M. Induces apoptosis. In intact neutrophils stimulated with fMet-Leu-Phe, LY294002 completely abolished PI 3-kinase activity at 50 M without cell toxicity.nn
nn
Specifications
nProvided as HCl saltnNot Sterilen
Featured in Publications
This item is non returnable