Learn More
Medchemexpress LLC LY294002 (hydrochloride) | 934389-88-5 | 99.9% | 369.8 g/mol | C19H17NO3·HCl | 5 MG
SDP
Catalog No. 5000213289
Encompass_Preferred
LY294002 hydrochloride is the hydrochloride salt of LY294002, a cell-permeable small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) and casein kinase 2 (CK2) used in in vitro studies of cell signaling, apoptosis, and autophagy. Supplied for research use with supporting documentation.
  • Inhibits PI3K and CK2 signaling pathways.
  • Suitable for in vitro cell signaling and apoptosis research.
  • High purity (99.9%).
  • Chemical formula C19H17NO3·HCl; molecular weight 369.8 g/mol.
  • Supplied in small research quantities (e.g., 5 mg).
  • Comes with SDS and certificate of analysis.
Catalog No. 50-002-13289 Supplier Medchemexpress LLC Supplier No. HY10108A5MG
LY294002 hydrochloride is the hydrochloride salt of LY294002, a cell-permeable small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) and casein kinase 2 (CK2) used in in vitro studies of cell signaling, apoptosis, and autophagy. Supplied for research use with supporting documentation.
  • Inhibits PI3K and CK2 signaling pathways.
  • Suitable for in vitro cell signaling and apoptosis research.
  • High purity (99.9%).
  • Chemical formula C19H17NO3·HCl; molecular weight 369.8 g/mol.
  • Supplied in small research quantities (e.g., 5 mg).
  • Comes with SDS and certificate of analysis.