Learn More
Medchemexpress LLC Ly294002 (hydrochloride) | 934389-88-5 | MFCD00270881 | 99.9% | 343.80 g/mol | C19H18ClNO3 | 100 MG
SDP
Catalog No. 5000213399
Encompass_Preferred
LY294002 hydrochloride is the hydrochloride salt of LY294002, a broad-spectrum phosphoinositide 3-kinase (PI3K) inhibitor that also shows activity against CK2 and DNA-PK. Supplied for research use in powder and solution formats, it is used to probe PI3K signaling, apoptosis, and related kinase pathways. CAS 934389-88-5; molecular weight 343.80 g/mol; high purity suitable for biochemical and cell-based assays.
  • Potent PI3K inhibitor useful for signaling studies.
  • Also inhibits CK2 and DNA-PK, enabling broader kinase pathway interrogation.
  • Available as powder and as DMSO solution for flexible dosing.
  • High purity suitable for biochemical and cell-based assays.
  • Stable when stored sealed at 4°C, away from moisture.
  • Widely used in oncology and cell signaling research applications.
Catalog No. 50-002-13399 Supplier Medchemexpress LLC Supplier No. HY10108A100MG
Add to Cart
Add to Cart
LY294002 hydrochloride is the hydrochloride salt of LY294002, a broad-spectrum phosphoinositide 3-kinase (PI3K) inhibitor that also shows activity against CK2 and DNA-PK. Supplied for research use in powder and solution formats, it is used to probe PI3K signaling, apoptosis, and related kinase pathways. CAS 934389-88-5; molecular weight 343.80 g/mol; high purity suitable for biochemical and cell-based assays.
  • Potent PI3K inhibitor useful for signaling studies.
  • Also inhibits CK2 and DNA-PK, enabling broader kinase pathway interrogation.
  • Available as powder and as DMSO solution for flexible dosing.
  • High purity suitable for biochemical and cell-based assays.
  • Stable when stored sealed at 4°C, away from moisture.
  • Widely used in oncology and cell signaling research applications.