Learn More
Medchemexpress LLC LY294002 hydrochloride | 934389-88-5 | MFCD01530034 | 99.9% | 343.8 g/mol | C19H18ClNO3 | 10 MG
SDP
Catalog No. 5000213345
Encompass_Preferred
LY294002 hydrochloride is a research-grade small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) and casein kinase 2 (CK2) used to investigate PI3K/AKT signaling, apoptosis, and autophagy in cell biology and cancer research. Provided as a solid reagent with high chemical purity for in vitro and biochemical studies; not for human or veterinary use.
  • Inhibits PI3K and CK2 kinase activity for pathway interrogation.
  • Supports studies of apoptosis, autophagy, and cell signaling.
  • High purity (99.93%) suitable for biochemical and cell-based assays.
  • Available in small solid quantities for laboratory workflows.
  • Standard chemical identifiers (CAS, MFCD) provided for reference.
Catalog No. 50-002-13345 Supplier Medchemexpress LLC Supplier No. HY10108A10MG
LY294002 hydrochloride is a research-grade small-molecule inhibitor of phosphoinositide 3-kinase (PI3K) and casein kinase 2 (CK2) used to investigate PI3K/AKT signaling, apoptosis, and autophagy in cell biology and cancer research. Provided as a solid reagent with high chemical purity for in vitro and biochemical studies; not for human or veterinary use.
  • Inhibits PI3K and CK2 kinase activity for pathway interrogation.
  • Supports studies of apoptosis, autophagy, and cell signaling.
  • High purity (99.93%) suitable for biochemical and cell-based assays.
  • Available in small solid quantities for laboratory workflows.
  • Standard chemical identifiers (CAS, MFCD) provided for reference.