Learn More
Medchemexpress LLC LY294002 (hydrochloride) | 934389-88-5 | MFCD01530034 | 99.9% | 343.8 g/mol | C19H18ClNO3 | 25 MG
SDP
Catalog No. 5000213034
Encompass_Preferred
LY294002 (hydrochloride) is a research-grade small-molecule inhibitor of phosphoinositide 3-kinases (PI3K) and casein kinase 2 (CK2), used to probe PI3K-mediated signaling, apoptosis, and autophagy in cellular and in vivo studies. It is supplied as a high-purity solid for laboratory research.
  • Potent inhibitor of PI3K and CK2.
  • High declared purity (>99.9%).
  • Supplied as a solid for laboratory use.
  • Molecular formula C19H18ClNO3; molecular weight 343.8 g/mol.
  • Available in small research quantities such as 25 MG.
  • Comes with an SDS and technical documentation for safe handling.
Catalog No. 50-002-13034 Supplier Medchemexpress LLC Supplier No. HY10108A25MG
LY294002 (hydrochloride) is a research-grade small-molecule inhibitor of phosphoinositide 3-kinases (PI3K) and casein kinase 2 (CK2), used to probe PI3K-mediated signaling, apoptosis, and autophagy in cellular and in vivo studies. It is supplied as a high-purity solid for laboratory research.
  • Potent inhibitor of PI3K and CK2.
  • High declared purity (>99.9%).
  • Supplied as a solid for laboratory use.
  • Molecular formula C19H18ClNO3; molecular weight 343.8 g/mol.
  • Available in small research quantities such as 25 MG.
  • Comes with an SDS and technical documentation for safe handling.