Learn More
Medchemexpress LLC LY294002 hydrochloride | 934389-88-5 | MFCD01530034 | 99.9% | 343.8 g/mol | C19H18ClNO3 | 50 MG
SDP
Catalog No. 5000211833
Encompass_Preferred
LY294002 hydrochloride is the hydrochloride salt of LY294002, a broad-spectrum phosphoinositide 3-kinase (PI3K) inhibitor supplied as a high-purity powder for research use only. It is commonly used in biochemical and cell-based studies to probe PI3K-dependent signaling, apoptosis, and autophagy pathways.
  • Broad-spectrum PI3K inhibitor, active against multiple PI3K isoforms.
  • Potent biochemical activity with submicromolar IC50 values for key isoforms.
  • High purity suitable for biochemical and cell-based assays (reported ~99.9%).
  • Hydrochloride salt form with improved solubility in polar solvents.
  • Supplied as a solid powder for convenient weighing and storage.
  • Commonly used to study apoptosis, autophagy, and cell signaling.
  • CAS number 934389-88-5 and molecular weight ≈343.8 g/mol.
  • Available in small research pack sizes including 50 MG.
Catalog No. 50-002-11833 Supplier Medchemexpress LLC Supplier No. HY10108A50MG
LY294002 hydrochloride is the hydrochloride salt of LY294002, a broad-spectrum phosphoinositide 3-kinase (PI3K) inhibitor supplied as a high-purity powder for research use only. It is commonly used in biochemical and cell-based studies to probe PI3K-dependent signaling, apoptosis, and autophagy pathways.
  • Broad-spectrum PI3K inhibitor, active against multiple PI3K isoforms.
  • Potent biochemical activity with submicromolar IC50 values for key isoforms.
  • High purity suitable for biochemical and cell-based assays (reported ~99.9%).
  • Hydrochloride salt form with improved solubility in polar solvents.
  • Supplied as a solid powder for convenient weighing and storage.
  • Commonly used to study apoptosis, autophagy, and cell signaling.
  • CAS number 934389-88-5 and molecular weight ≈343.8 g/mol.
  • Available in small research pack sizes including 50 MG.