Learn More
TARGETMOL CHEMICALS INC LY2940094 5MG
SDP
Catalog No. 502581201
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY2940094 (LY-2940094) decreases ethanol self-administration in animal models. LY2940094 is an effective selective and orally available nociceptin receptor antagonist with high affinity (Ki=0.105 nM) and antagonist potency (Kb=0.166 nM). purity: 99%
Catalog No. 50-258-1201 Supplier TARGETMOL CHEMICALS INC Supplier No. T157995MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY2940094 (LY-2940094) decreases ethanol self-administration in animal models. LY2940094 is an effective selective and orally available nociceptin receptor antagonist with high affinity (Ki=0.105 nM) and antagonist potency (Kb=0.166 nM). purity: 99%