Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY2940094 tartrate 10mg | 1307245-87-9 | 10 MG
SDP

Catalog No. 5000229588
Encompass_Preferred

LY2940094 tartrate is the tartrate salt of a selective nociceptin (NOP) receptor antagonist used in preclinical pharmacology and behavioral research. It is supplied as a white to off-white solid with high reported purity and characterized molecular properties suitable for analytical and dosing studies.

  • Tartrate salt form, solid (white to off-white).
  • High purity by LCMS (reported ~99.66%).
  • Molecular weight 631.05 g/mol; formula C26H29ClF2N4O8S.
  • Intended for preclinical pharmacology and behavioral studies.
  • Available in small milligram-scale packages for analytical use and dosing.

Catalog No. 50-002-29588 Supplier Medchemexpress LLC Supplier No. HY114452A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY2940094 tartrate is the tartrate salt of a selective nociceptin (NOP) receptor antagonist used in preclinical pharmacology and behavioral research. It is supplied as a white to off-white solid with high reported purity and characterized molecular properties suitable for analytical and dosing studies.

  • Tartrate salt form, solid (white to off-white).
  • High purity by LCMS (reported ~99.66%).
  • Molecular weight 631.05 g/mol; formula C26H29ClF2N4O8S.
  • Intended for preclinical pharmacology and behavioral studies.
  • Available in small milligram-scale packages for analytical use and dosing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.