Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY2940094 tartrate 10mM 1mL | 1307245-87-9 | 1 ML
SDP

Catalog No. 5000228503
Encompass_Preferred

LY2940094 tartrate is a potent, brain-penetrant nociceptin (NOP) receptor antagonist supplied as a 10 mM solution in DMSO for laboratory research use.

  • 10 mM solution in DMSO
  • 1 mL vial
  • Purity 99.7%
  • Tartrate salt form
  • CAS 1307245-87-9
  • Molecular weight 631.05 g/mol
  • For research use only

Catalog No. 50-002-28503 Supplier Medchemexpress LLC Supplier No. HY114452A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY2940094 tartrate is a potent, brain-penetrant nociceptin (NOP) receptor antagonist supplied as a 10 mM solution in DMSO for laboratory research use.

  • 10 mM solution in DMSO
  • 1 mL vial
  • Purity 99.7%
  • Tartrate salt form
  • CAS 1307245-87-9
  • Molecular weight 631.05 g/mol
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.