Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY2940094 tartrate 50mg | 1307245-87-9 | 50 MG
SDP

Catalog No. 5000228500
Encompass_Preferred

LY2940094 tartrate is the tartrate salt of LY2940094, a potent, brain-penetrant nociceptin (NOP) receptor antagonist supplied as a solid for research use.

  • Supplied as a 50 mg solid powder.
  • High purity suitable for in vitro and preclinical studies.
  • Molecular weight 631.05 g/mol and formula C26H29ClF2N4O8S.
  • Store sealed at 4°C; in solution store at -80°C for long-term storage.
  • Provided with a batch-specific certificate of analysis.

Catalog No. 50-002-28500 Supplier Medchemexpress LLC Supplier No. HY114452A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY2940094 tartrate is the tartrate salt of LY2940094, a potent, brain-penetrant nociceptin (NOP) receptor antagonist supplied as a solid for research use.

  • Supplied as a 50 mg solid powder.
  • High purity suitable for in vitro and preclinical studies.
  • Molecular weight 631.05 g/mol and formula C26H29ClF2N4O8S.
  • Store sealed at 4°C; in solution store at -80°C for long-term storage.
  • Provided with a batch-specific certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.