Learn More
Apexbio Technology LLC LY2940680 1258861-20-9 200mg
SDP
Catalog No. 5000061729
Encompass_Preferred
LY2940680 (CAS 1258861-20-9) is a selective antagonist of the Smoothened (Smo) receptor a seven-transmembrane G protein-coupled receptor involved in regulating Hedgehog (Hh) signalling LY2940680 binds primarily to the extracellular loops of Smo distinguishing it from previously described inhibitors such as SANT-1 which bind to the transmembrane domain By blocking Smo function LY2940680 suppresses downstream Hh pathway activation resulting in reduced cell proliferation observed in cell lines with Smo mutations or resistance to vismodegib Investigations of LY2940680 in basal cell carcinoma and medulloblastoma highlight its potential utility as a single-agent therapeutic targeting cancer stem cell populations
Catalog No. 50-000-61729 Supplier Apexbio Technology LLC Supplier No. A8651200
LY2940680 (CAS 1258861-20-9) is a selective antagonist of the Smoothened (Smo) receptor a seven-transmembrane G protein-coupled receptor involved in regulating Hedgehog (Hh) signalling LY2940680 binds primarily to the extracellular loops of Smo distinguishing it from previously described inhibitors such as SANT-1 which bind to the transmembrane domain By blocking Smo function LY2940680 suppresses downstream Hh pathway activation resulting in reduced cell proliferation observed in cell lines with Smo mutations or resistance to vismodegib Investigations of LY2940680 in basal cell carcinoma and medulloblastoma highlight its potential utility as a single-agent therapeutic targeting cancer stem cell populations