Learn More
Abcam LY2940680 (Taladegib), 50MG
Catalog No. 502672008
Encompass_Preferred
MW 512.5 Da. Potent Hedgehog signaling pathway inhibitor. Smoothened antagonist. Inhibits growth of cancer cell lines containing a disease-relevant Smoothened gene mutation. Has anti-tumor activity in animal models.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2008 Supplier Abcam Supplier No. AB26987450MG
MW 512.5 Da. Potent Hedgehog signaling pathway inhibitor. Smoothened antagonist. Inhibits growth of cancer cell lines containing a disease-relevant Smoothened gene mutation. Has anti-tumor activity in animal models.
The product is subject to the following: Abcam Restricted Use Statement