Learn More
Medchemexpress LLC HY-107765 5mg Medchemexpress, LY2955303 CAS:1433497-19-8 Purity:>98%
SDP
Catalog No. 501968462
Encompass_Preferred
Medchemexpress, HY-107765 5mg LY2955303 CAS:1433497-19-8 LY2955303 is a potent and selective retinoic acid receptor gamma (RARγ) antagonist with a Ki of 1.09 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8462 Supplier Medchemexpress LLC Supplier No. HY1077655MG
Medchemexpress, HY-107765 5mg LY2955303 CAS:1433497-19-8 LY2955303 is a potent and selective retinoic acid receptor gamma (RARγ) antagonist with a Ki of 1.09 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.