Learn More
Apexbio Technology LLC LY3009120 1454682-72-4 25mg
SDP
Catalog No. 5000096090
Encompass_Preferred
LY3009120 (CAS 1454682-72-4) is a pan-RAF inhibitor that targets both monomeric and dimeric forms of RAF kinases including wild-type BRAF CRAF and mutant forms such as BRAFV600E and BRAFV600E G468A with IC50 values ranging from 5 8 to 17 nM By suppressing RAF activity LY3009120 disrupts downstream phosphorylation of MEK and ERK modulating MAPK/ERK signaling involved in cell proliferation and survival In cell models harboring NRAS or KRAS mutations as well as BRAF V600E mutant melanoma LY3009120 induces G0/G1 cell cycle arrest and minimizes paradoxical pathway activation In vivo it achieves dose-dependent inhibition of MEK and ERK phosphorylation and drives near-complete tumor regression in xenograft models LY3009120 is valuable for investigating RAF-mediated oncogenic processes and resistance mechanisms in MAPK-driven cancers
Catalog No. 50-000-96090 Supplier Apexbio Technology LLC Supplier No. A871625
LY3009120 (CAS 1454682-72-4) is a pan-RAF inhibitor that targets both monomeric and dimeric forms of RAF kinases including wild-type BRAF CRAF and mutant forms such as BRAFV600E and BRAFV600E G468A with IC50 values ranging from 5 8 to 17 nM By suppressing RAF activity LY3009120 disrupts downstream phosphorylation of MEK and ERK modulating MAPK/ERK signaling involved in cell proliferation and survival In cell models harboring NRAS or KRAS mutations as well as BRAF V600E mutant melanoma LY3009120 induces G0/G1 cell cycle arrest and minimizes paradoxical pathway activation In vivo it achieves dose-dependent inhibition of MEK and ERK phosphorylation and drives near-complete tumor regression in xenograft models LY3009120 is valuable for investigating RAF-mediated oncogenic processes and resistance mechanisms in MAPK-driven cancers