Learn More
Medchemexpress LLC HY-12558 100mg , LY3009120 DP-4978 CAS:1454682-72-4 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY12558100MG
Encompass_Preferred
Medchemexpress, HY-12558 100mg LY3009120 DP-4978 CAS:1454682-72-4 Formula:C23H29FN6O IC50: 5.8 nM (BRAF V600E ), 9.1 nM (BRAF WT ), 15 nM (CRAF WT ) Purity:98% LY3009120 is a pan RAF and RAF dimer inhibitor, and inhibits BRAF V600E , BRAF WT and CRAF WT with IC 50 values of 5.8, 9.1 and 15 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2950