Learn More
Medchemexpress LLC HY-108707 5mg Medchemexpress, LY3130481 CAS:1610802-47-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1087075MG
Encompass_Preferred
Medchemexpress, HY-108707 5mg LY3130481 CAS:1610802-47-5 LY3130481 is an AMPA receptor antagonist that is dependent upon transmembrane AMPA receptor regulatory protein (TARP) γ-8, selective inhibits AMPA/TARP γ-8 with an IC50 of 65 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9644