Learn More
Medchemexpress LLC HY-100023 5mg Medchemexpress, LY3177833 CAS:1627696-51-8 Purity:>98%
SDP
Catalog No. 501931695
Encompass_Preferred
Medchemexpress, HY-100023 5mg LY3177833 CAS:1627696-51-8 LY3177833 is a CDC7 and pMCM2 inhibitor with IC 50 values of 3.3 nM and 290 nM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1695 Supplier Medchemexpress LLC Supplier No. HY1000235MG
Medchemexpress, HY-100023 5mg LY3177833 CAS:1627696-51-8 LY3177833 is a CDC7 and pMCM2 inhibitor with IC 50 values of 3.3 nM and 290 nM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.