Learn More
Medchemexpress LLC HY-101494 1mg Medchemexpress, LY3214996 CAS:1951483-29-6 Purity:>98%
SDP
Catalog No. 501932152
Encompass_Preferred
Medchemexpress, HY-101494 1mg LY3214996 CAS:1951483-29-6 LY3214996 is a highly selective inhibitor of ERK1 and ERK2 , with IC 50 of 5 nM for both enzymes in biochemical assays. LY3214996 potently inhibits cellular p-RSK1 in BRAF and RAS mutant cancer cell lines. LY3214996 shows potent antitumor activities in cancer models with MAPK pathway alterations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2152 Supplier Medchemexpress LLC Supplier No. HY1014941MG
Medchemexpress, HY-101494 1mg LY3214996 CAS:1951483-29-6 LY3214996 is a highly selective inhibitor of ERK1 and ERK2 , with IC 50 of 5 nM for both enzymes in biochemical assays. LY3214996 potently inhibits cellular p-RSK1 in BRAF and RAS mutant cancer cell lines. LY3214996 shows potent antitumor activities in cancer models with MAPK pathway alterations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.