Learn More
Medchemexpress LLC HY-114258 10mg Medchemexpress, LY3295668 CAS:1919888-06-4 Purity:>98%
SDP
Catalog No. 501969314
Encompass_Preferred
Medchemexpress, HY-114258 10mg LY3295668 CAS:1919888-06-4 LY3295668 is a potent, orally active and highly specific Aurora-A kinase inhibitor, with Ki values of 0.8 nM and 1038 nM for AurA and AurB, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9314 Supplier Medchemexpress LLC Supplier No. HY11425810MG
Medchemexpress, HY-114258 10mg LY3295668 CAS:1919888-06-4 LY3295668 is a potent, orally active and highly specific Aurora-A kinase inhibitor, with Ki values of 0.8 nM and 1038 nM for AurA and AurB, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.