Learn More
Chemie Tek LY3295668(LY-3295668, AK-5MG
SDP
Catalog No. NC2009954
Encompass_Preferred
LY3295668 (LY-3295668, AK-01), Aurora A kinase (AurA) inhibitor with Ki of 0.8 nM, and H446 AurA auto-P IC50 of 0.6 nM, highly selective (>1000 fold) over Aurora B. LY3295668 is cytotoxic to RB1 mutant cancer cells (NCI-H446 cell IC50 = 0.752 uM), 5MG
Catalog No. NC2009954 Supplier Chemie Tek Supplier No. CTLY32955MG
LY3295668 (LY-3295668, AK-01), Aurora A kinase (AurA) inhibitor with Ki of 0.8 nM, and H446 AurA auto-P IC50 of 0.6 nM, highly selective (>1000 fold) over Aurora B. LY3295668 is cytotoxic to RB1 mutant cancer cells (NCI-H446 cell IC50 = 0.752 uM), 5MG