Learn More
Selleck Chemical LLC Ruboxistaurin (LY333531) HCl 2mg 169939-93-9 Ruboxistaurin hydrochloride, LY333531 hydrochloride
SDP
Catalog No. 501365093
Encompass_Preferred
Ruboxistaurin (LY333531) HCl is a ?-specific protein kinase C inhibitor. It competitively and reversibly inhibits PKC?1 and PKC?2 with IC50 values of 4.7 and 5.9 nM respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5093 Supplier Selleck Chemical LLC Supplier No. S76632MG
Ruboxistaurin (LY333531) HCl is a ?-specific protein kinase C inhibitor. It competitively and reversibly inhibits PKC?1 and PKC?2 with IC50 values of 4.7 and 5.9 nM respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.