Learn More
Medchemexpress LLC HY-103107 5mg Medchemexpress, LY334370 CAS:182563-08-2 Purity:>98%
SDP
Catalog No. 501968038
Encompass_Preferred
Medchemexpress, HY-103107 5mg LY334370 CAS:182563-08-2 LY334370 is a selective 5-HT1F receptor agonist with a Ki of 1.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8038 Supplier Medchemexpress LLC Supplier No. HY1031075MG
Medchemexpress, HY-103107 5mg LY334370 CAS:182563-08-2 LY334370 is a selective 5-HT1F receptor agonist with a Ki of 1.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.