Learn More
Selleck Chemical LLC Zosuquidar (LY335979) 3HCl 5mg 167465-36-3 RS 33295-198 (D06387) 3HCl
SDP
Supplier:  Selleck Chemical LLC S14815MG
Encompass_Preferred
Zosuquidar 3HCl (LY335979, RS 33295-198, D06387) is a potent modulator of P-glycoprotein-mediated multi-drug resistance with Ki of 60 nM in a cell-free assay. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0291877