Learn More
TARGETMOL CHEMICALS INC LY3372689 5MG
Supplier: TARGETMOL CHEMICALS INC T616615MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY3372689 (Formulaic Ia) is an orally active O-GlcNAcase (OGA) enzyme inhibitor for tau protein-based disorders including Alzheimer's disease. purity: 99%