Learn More
TARGETMOL CHEMICALS INC LY339434 HCL 5MG
SDP
Catalog No. 502580557
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LY339434 HCl is a low affinity GluR5 erythrocyte receptor agonist.LY339434 HCl causes rapid neuronal death primarily by affecting N-methyl-D-aspartate (NMDA) receptors.
Catalog No. 50-258-0557 Supplier TARGETMOL CHEMICALS INC Supplier No. T27947L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LY339434 HCl is a low affinity GluR5 erythrocyte receptor agonist.LY339434 HCl causes rapid neuronal death primarily by affecting N-methyl-D-aspartate (NMDA) receptors.