Learn More
Selleck Chemical LLC LY364947 10mg 396129-53-6 HTS 466284
SDP
Supplier:  Selleck Chemical LLC S280510MG
Encompass_Preferred
LY364947 (HTS 466284) is a potent ATP-competitive inhibitor of TGF?R-I with IC50 of 59 nM in a cell-free assay, shows 7-fold selectivity over TGF?R-II. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2989