Learn More
Apexbio Technology LLC LY364947, 10mg. Cas: 396129-53-6 MFCD: MFCD00800758. inhibitor of TGF-β type I receptor kinase domain
SDP
Supplier:  Apexbio Technology LLC B228710
Encompass_Preferred
LY364947 is a potent ATP-competitive inhibitor of TGFBR-I with IC50 of 59 nM, shows 7-fold selectivity over TGFBR-II. 396129-53-6. MFCD00800758
Catalog No. 50-101-1321