Learn More
Apexbio Technology LLC LY364947 396129-53-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062438
Encompass_Preferred
LY364947 (CAS 396129-53-6) is a selective inhibitor targeting the kinase domain of transforming growth factor- (TGF- ) type I receptor TGF- signaling regulates crucial biological processes such as cell proliferation differentiation and tissue homeostasis In vitro studies demonstrated that LY364947 inhibits the receptor kinase activity with an IC50 of 51 nM effectively suppressing TGF- -dependent luciferase activity in mink lung epithelial cells (p3TP Lux assay) and growth of NIH 3T3 fibroblasts In vivo LY364947 showed protective effects against NMDA-induced retinal degeneration in rats attenuating retinal ganglion cell loss and capillary degradation Thus LY364947 serves as a valuable pharmacological tool in TGF- -related research and associated pathological conditions
Catalog No. 50-000-62438 Supplier Apexbio Technology LLC Supplier No. B22875.1
LY364947 (CAS 396129-53-6) is a selective inhibitor targeting the kinase domain of transforming growth factor- (TGF- ) type I receptor TGF- signaling regulates crucial biological processes such as cell proliferation differentiation and tissue homeostasis In vitro studies demonstrated that LY364947 inhibits the receptor kinase activity with an IC50 of 51 nM effectively suppressing TGF- -dependent luciferase activity in mink lung epithelial cells (p3TP Lux assay) and growth of NIH 3T3 fibroblasts In vivo LY364947 showed protective effects against NMDA-induced retinal degeneration in rats attenuating retinal ganglion cell loss and capillary degradation Thus LY364947 serves as a valuable pharmacological tool in TGF- -related research and associated pathological conditions