Learn More
Medchemexpress LLC Ly367385 hydrochloride | 2829282-00-8 | 99.4% | 245.66 | C10H12ClNO4 | 100 MG
SDP
Catalog No. 5000239790
Encompass_Preferred
LY367385 hydrochloride is a selective and potent antagonist of the metabotropic glutamate receptor mGluR1a used in research to investigate mGluR1-mediated signaling, neuroprotective mechanisms, and seizure models. The compound is supplied as a solid suitable for laboratory handling and storage.
  • Selective mGluR1a antagonist for receptor-specific studies.
  • High reported purity supports reproducible results.
  • Solid, stable form suitable for standard laboratory storage.
  • Applicable to neuroprotection, anticonvulsant, and antiepileptic research.
  • Compatible with common solvent storage conditions for short- and long-term use.
Catalog No. 50-002-39790 Supplier Medchemexpress LLC Supplier No. HY107515A100MG
LY367385 hydrochloride is a selective and potent antagonist of the metabotropic glutamate receptor mGluR1a used in research to investigate mGluR1-mediated signaling, neuroprotective mechanisms, and seizure models. The compound is supplied as a solid suitable for laboratory handling and storage.
  • Selective mGluR1a antagonist for receptor-specific studies.
  • High reported purity supports reproducible results.
  • Solid, stable form suitable for standard laboratory storage.
  • Applicable to neuroprotection, anticonvulsant, and antiepileptic research.
  • Compatible with common solvent storage conditions for short- and long-term use.