Learn More
Medchemexpress LLC Ly367385 hydrochloride | 2829282-00-8 | 99.4% | 245.66 g/mol | C10H12ClNO4 | 10 MG
SDP
Catalog No. 5000239812
Encompass_Preferred
LY367385 hydrochloride is the hydrochloride salt of a selective mGluR1a antagonist intended for research use. Supplied as a high-purity solid, it is suitable for in vitro and preclinical studies; store sealed and protected from moisture.
Catalog No. 50-002-39812 Supplier Medchemexpress LLC Supplier No. HY107515A10MG
LY367385 hydrochloride is the hydrochloride salt of a selective mGluR1a antagonist intended for research use. Supplied as a high-purity solid, it is suitable for in vitro and preclinical studies; store sealed and protected from moisture.