Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LY367385 hydrochloride | 2829282-00-8 | 99.4% | 245.66 g·mol⁻¹ | C10H12ClNO4 | 5 MG
SDP

Catalog No. 5000239677
Encompass_Preferred

LY367385 hydrochloride is a selective metabotropic glutamate receptor 1a (mGluR1a) antagonist used as a research reagent in neuropharmacology. It inhibits quisqualate-induced phosphoinositide hydrolysis (IC50 = 8.8 μM) and is provided as a solid for in vitro and formulation studies.

  • High purity: 99.44%.
  • Molecular weight: 245.66 g·mol⁻¹.
  • CAS number: 2829282-00-8.
  • Soluble in DMSO (125 mg/mL) and water (12.5 mg/mL) with ultrasonic aid.
  • In vivo formulation protocols produce clear solutions ≥2.08 mg/mL in common vehicles.
  • Storage: solid at 4°C; stock solutions at -80°C (6 months) or -20°C (1 month).
  • Available in small-scale packages suitable for research, including 5 mg.

Catalog No. 50-002-39677 Supplier Medchemexpress LLC Supplier No. HY107515A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY367385 hydrochloride is a selective metabotropic glutamate receptor 1a (mGluR1a) antagonist used as a research reagent in neuropharmacology. It inhibits quisqualate-induced phosphoinositide hydrolysis (IC50 = 8.8 μM) and is provided as a solid for in vitro and formulation studies.

  • High purity: 99.44%.
  • Molecular weight: 245.66 g·mol⁻¹.
  • CAS number: 2829282-00-8.
  • Soluble in DMSO (125 mg/mL) and water (12.5 mg/mL) with ultrasonic aid.
  • In vivo formulation protocols produce clear solutions ≥2.08 mg/mL in common vehicles.
  • Storage: solid at 4°C; stock solutions at -80°C (6 months) or -20°C (1 month).
  • Available in small-scale packages suitable for research, including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.