Learn More
TARGETMOL CHEMICALS INC LY404039 25MG
SDP
Catalog No. 502579633
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY404039 (pomaglumetad) is an effective agonist of recombinant human mGlu2(Ki =149 nM)/mGlu3(Ki =92 nM). purity: 99%
Catalog No. 50-257-9633 Supplier TARGETMOL CHEMICALS INC Supplier No. T602225MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY404039 (pomaglumetad) is an effective agonist of recombinant human mGlu2(Ki =149 nM)/mGlu3(Ki =92 nM). purity: 99%