Learn More
Selleck Chemical LLC LY411575 5mg 209984-57-6
SDP
Supplier:  Selleck Chemical LLC S27145MG
Encompass_Preferred
LY411575 is a potent ?-secretase inhibitor with IC50 of 0.078 nM/0.082 nM (membrane/cell-based), also inhibits Notch cleavage with IC50 of 0.39 nM in APP or N?E expressing HEK293 cells. LY411575 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0960467