Learn More
Cayman Chemical LY456236 (hydrochloride), Purity: ≥98%, CAS: 338736-46-2, 10 mg
SDP
Catalog No. 502943685
Encompass_Preferred
An antagonist of mGluR1 (IC50 = 140 nM); reduces the growth of PC-9 cells and PC-9-AMG cells in MGS co-culture but not in monoculture; reduces MGS co-culture-induced increases in EGFR levels in PC-9-AMG cells; reduces tumor growth in a PC-9-AMG mouse model of brain metastasis at 50 mg/kg per day; inhibits seizures in a variety of seizure models, including inhibition of audiogenic clonic-tonic seizures in mice (ED50 = 35 mg/kg); reduces second-phase, but not first-phase, formalin-induced hyperalgesia at 30 and 100 mg/kg
Catalog No. 50-294-3685 Supplier Cayman Chemical Supplier No. 4226310 MG
An antagonist of mGluR1 (IC50 = 140 nM); reduces the growth of PC-9 cells and PC-9-AMG cells in MGS co-culture but not in monoculture; reduces MGS co-culture-induced increases in EGFR levels in PC-9-AMG cells; reduces tumor growth in a PC-9-AMG mouse model of brain metastasis at 50 mg/kg per day; inhibits seizures in a variety of seizure models, including inhibition of audiogenic clonic-tonic seizures in mice (ED50 = 35 mg/kg); reduces second-phase, but not first-phase, formalin-induced hyperalgesia at 30 and 100 mg/kg