Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Ly6k (E3M7W) Rabbit Monoclonal Antibody, Mouse, Unconjugated, 100 μL
SDP

Catalog No. 502730454
Encompass

Ly6k (E3M7W) Rabbit Monoclonal Antibody is a recombinant antibody that recognizes endogenous levels of total Ly6k protein. This monoclonal antibody is generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro70 of murine Ly6k protein. It is provided in a buffer of 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.

  • Host species: Rabbit
  • Reactivity: Mouse
  • Validated applications: Western blotting, immunoprecipitation, and immunofluorescence (frozen)
  • Recommended dilutions: 1:1000 for western blotting, 1:50 for immunoprecipitation, and 1:50 to 1:100 for immunofluorescence (frozen)
  • Molecular weight: 18 kDa
  • Storage: -20°C

Catalog No. 50-273-0454 Supplier Cell Signaling Technology Supplier No. 87994S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ly6k (E3M7W) Rabbit Monoclonal Antibody is a recombinant antibody that recognizes endogenous levels of total Ly6k protein. This monoclonal antibody is generated by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro70 of murine Ly6k protein. It is provided in a buffer of 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.

  • Host species: Rabbit
  • Reactivity: Mouse
  • Validated applications: Western blotting, immunoprecipitation, and immunofluorescence (frozen)
  • Recommended dilutions: 1:1000 for western blotting, 1:50 for immunoprecipitation, and 1:50 to 1:100 for immunofluorescence (frozen)
  • Molecular weight: 18 kDa
  • Storage: -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.