Learn More
TARGETMOL CHEMICALS INC LY88074 5MG
SDP
Catalog No. 502581502
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY88074 is a raloxifene analog in which the basic side chain has been removed. LY88074 is an agonist of ER(beta) (EC50 = 232 nM).This compound stimulates uterine cell proliferation. purity: 98%
Catalog No. 50-258-1502 Supplier TARGETMOL CHEMICALS INC Supplier No. T119155MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY88074 is a raloxifene analog in which the basic side chain has been removed. LY88074 is an agonist of ER(beta) (EC50 = 232 nM).This compound stimulates uterine cell proliferation. purity: 98%