Learn More
Medchemexpress LLC HY-114307 5mg Medchemexpress, Ly93 CAS:1883528-69-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1143075MG
Encompass_Preferred
Medchemexpress, HY-114307 5mg Ly93 CAS:1883528-69-5 Ly93 is a selective and orally active sphingomyelin synthase 2 (SMS2) inhibitor, with an IC50 of 91 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9784