Learn More
Aviva Systems Biology Corp LY96 Antibody - C-terminal region (ARP61217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LY96. It was validated on Western Blot. . Alternate names: MD-2, MD2, ly-96, ESOP-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-299