Learn More
Aviva Systems Biology Corp Lyar Antibody - N-terminal region (ARP57041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lyar. It was validated on Western Blot. . Alternate names: MLZ-264ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-257