Learn More
Aviva Systems Biology Corp LYCAT Antibody - middle region (ARP44426_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44426_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LYCAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LYCAT. Alternate names: ALCAT1, FLJ37965, UNQ1849, LYCAT, AGPAT8, 1AGPAT8, HSRG1849ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53421