Learn More
Cayman Chemical Lycopene, 502-65-8, 1 mg
SDP
Supplier:  Cayman Chemical 709451
Encompass_Preferred
A red-colored carotenoid found in tomatoes and other red fruits and vegetables; powerful antioxidant that efficiently quenches singlet oxygen
Catalog No. NC1142173