Learn More
Medchemexpress LLC HY-N0287 10mg , Lycopene CAS:502-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN028710MG
Encompass_Preferred
HY-N0287 10mg Lycopene CAS: 502-65-8 Lycopene is naturally occurring carotenoids found in tomato, tomato products, and in other red fruits and vegetables; exhibits antioxidant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9824